Gene Transcripts

Gene Symbol
PSMG4
Entrez Gene ID
389362
Gene Name
proteasome assembly chaperone 4
Gene Aliases
C6orf86, PAC4, bA506K6.2
Chromosome Location
Chr. 6: 3254325 - 3308224 on Build GRCh38
UniGene ID
Hs.207069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK096543.1 - 1 1631

View Full Product Details