Gene Transcripts
- Gene Symbol
- RPL9P11
- Entrez Gene ID
- 642740
- Gene Name
- ribosomal protein L9 pseudogene 11
- Gene Aliases
- RPL9_1_9
- Chromosome Location
- Chr. 1: 9576405 - 9577081 on Build GRCh38
- UniGene ID
- Hs.661110
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130277.1 |
- |
1 |
433 |
|
View Full Product Details