Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005267510.1 | XP_005267567.1 | 7 | 4306 | |
GenBank mRNA | AK025898.1 | - | 1 | 918 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM917307.1 | - | 116 | Chr. - - [-] |