Gene Transcripts
- Gene Symbol
- TCTN1
- Entrez Gene ID
- 79600
- Gene Name
- tectonic family member 1
- Gene Aliases
- JBTS13, TECT1
- Chromosome Location
- Chr. 12: 110614027 - 110649130 on Build GRCh38
- UniGene ID
- Hs.211511
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC030993.1 |
- |
1 |
485 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG399158.1 |
- |
1 |
483 |
|
Chr. 12 - 110632450 - 110633076 [+] |
View Full Product Details