Gene Transcripts
- Gene Symbol
- NANP
- Entrez Gene ID
- 140838
- Gene Name
- N-acetylneuraminic acid phosphatase
- Gene Aliases
- C20orf147, HDHD4, dJ694B14.3
- Chromosome Location
- Chr. 20: 25612935 - 25624012 on Build GRCh38
- UniGene ID
- Hs.143137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074335.1 |
- |
1 |
1712 |
|
View Full Product Details