Gene Transcripts

Gene Symbol
SUGT1
Entrez Gene ID
10910
Gene Name
SGT1 homolog, MIS12 kinetochore complex assembly cochaperone
Gene Aliases
SGT1
Chromosome Location
Chr. 13: 52652691 - 52700756 on Build GRCh38
UniGene ID
Hs.281902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011534891.2 XP_011533193.1 14 2071
XM_011534893.2 XP_011533195.1 13 1789
XM_017020365.1 XP_016875854.1 14 1961
XM_017020366.1 XP_016875855.1 14 1836
XM_017020367.1 XP_016875856.1 14 1870
XM_017020368.1 XP_016875857.1 14 1882


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR616065.1 - 1 1340 Chr. 13 - 52687144 - 52689945 [+]

View Full Product Details