Gene Transcripts
- Gene Symbol
- KANK2
- Entrez Gene ID
- 25959
- Gene Name
- KN motif and ankyrin repeat domains 2
- Gene Aliases
- ANKRD25, MXRA3, PPKWH, SIP
- Chromosome Location
- Chr. 19: 11164267 - 11197567 on Build GRCh38
- UniGene ID
- Hs.284208
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC019078.2 |
- |
1 |
589 |
|
View Full Product Details