Gene Transcripts

Gene Symbol
CMIP
Entrez Gene ID
80790
Gene Name
c-Maf inducing protein
Gene Aliases
TCMIP
Chromosome Location
Chr. 16: 81445170 - 81711762 on Build GRCh38
UniGene ID
Hs.594095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017023733.1 XP_016879222.1 1 549
GenBank mRNA AK127550.1 - 1 549

View Full Product Details