Gene Transcripts
- Gene Symbol
- OK/SW-CL.58
- Entrez Gene ID
- 100131766
- Gene Name
- OK/SW-CL.58
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.683432
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB064667.1 |
- |
1 |
1633 |
|
View Full Product Details