Gene Transcripts
- Gene Symbol
- EDN1
- Entrez Gene ID
- 1906
- Gene Name
- endothelin 1
- Gene Aliases
- ARCND3, ET1, HDLCQ7, PPET1, QME
- Chromosome Location
- Chr. 6: 12256464 - 12297194 on Build GRCh38
- UniGene ID
- Hs.511899
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036851.1 |
- |
4 |
795 |
|
BM680754.1 |
- |
1 |
349 |
|
BQ009676.1 |
- |
1 |
349 |
|
View Full Product Details