Gene Transcripts

Gene Symbol
PAWR
Entrez Gene ID
5074
Gene Name
pro-apoptotic WT1 regulator
Gene Aliases
PAR4, Par-4
Chromosome Location
Chr. 12: 79587791 - 79691097 on Build GRCh38
UniGene ID
Hs.643130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006719436.3 XP_006719499.1 5 3868
GenBank mRNA AK027848.2 - 1 1369


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CA395056.1 - 1 479 Chr. 12 - 79601867 - 79602388 [-]

View Full Product Details