Gene Transcripts

Gene Symbol
C18orf63
Entrez Gene ID
644041
Gene Name
chromosome 18 open reading frame 63
Gene Aliases
DKFZP781G0119
Chromosome Location
Chr. 18: 74315875 - 74359187 on Build GRCh38
UniGene ID
Hs.465414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174123.1 NP_001167594.1 14 3539
GenBank mRNA CR749350.1 - 14 3539

View Full Product Details