Gene Transcripts

Gene Symbol
RHOBTB2
Entrez Gene ID
23221
Gene Name
Rho related BTB domain containing 2
Gene Aliases
DBC2
Chromosome Location
Chr. 8: 22987406 - 23020197 on Build GRCh38
UniGene ID
Hs.372688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017013251.1 XP_016868740.1 1 2055
GenBank mRNA AK092617.1 - 1 2055

View Full Product Details