Gene Transcripts

Gene Symbol
DNAJB14
Entrez Gene ID
79982
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Gene Aliases
EGNR9427, PRO34683
Chromosome Location
Chr. 4: 99896248 - 99946726 on Build GRCh38
UniGene ID
Hs.577426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031723.3 NP_001026893.1 8 5580
NM_001278310.1 NP_001265239.1 7 5408
GenBank mRNA AK095074.1 - 1 1664

View Full Product Details