Gene Transcripts
- Gene Symbol
- TTC1
- Entrez Gene ID
- 7265
- Gene Name
- tetratricopeptide repeat domain 1
- Gene Aliases
- TPR1
- Chromosome Location
- Chr. 5: 160009100 - 160065545 on Build GRCh38
- UniGene ID
- Hs.519718
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128840.1 |
- |
1 |
830 |
|
View Full Product Details