Gene Transcripts
- Gene Symbol
- SLC38A9
- Entrez Gene ID
- 153129
- Gene Name
- solute carrier family 38 member 9
- Gene Aliases
- URLC11
- Chromosome Location
- Chr. 5: 55625845 - 55712344 on Build GRCh38
- UniGene ID
- Hs.742101
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_047649.1 |
|
12 |
1735 |
|
GenBank mRNA |
BX641065.1 |
- |
6 |
2606 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK057666.1 |
- |
1 |
1377 |
|
Chr. 5 - 55633021 - 55635083 [-] |
View Full Product Details