Gene Transcripts

Gene Symbol
TFDP2
Entrez Gene ID
7029
Gene Name
transcription factor Dp-2
Gene Aliases
DP2
Chromosome Location
Chr. 3: 141944428 - 142149544 on Build GRCh38
UniGene ID
Hs.379018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178138.1 NP_001171609.1 14 7054
NM_001178139.1 NP_001171610.1 13 7016
NM_001178140.1 NP_001171611.1 10 6512
NM_001178141.1 NP_001171612.1 10 6445
NM_001178142.1 NP_001171613.1 8 6293
NM_006286.4 NP_006277.1 11 6570
XM_005247731.2 XP_005247788.1 13 7019
XM_011513101.2 XP_011511403.1 14 7115
XM_011513102.2 XP_011511404.1 12 6688
XM_011513106.2 XP_011511408.1 14 7112
XM_011513108.2 XP_011511410.1 11 6842
XM_011513109.2 XP_011511411.1 9 6414
XM_011513111.2 XP_011511413.1 9 6411
XM_017007091.1 XP_016862580.1 13 7027
XM_017007092.1 XP_016862581.1 16 7273
XM_017007093.1 XP_016862582.1 16 7462
XM_017007094.1 XP_016862583.1 13 7048
XM_017007095.1 XP_016862584.1 15 7238
XM_017007096.1 XP_016862585.1 12 6945
XM_017007097.1 XP_016862586.1 14 7044
XM_017007098.1 XP_016862587.1 14 7192
XM_017007099.1 XP_016862588.1 15 7167
XM_017007101.1 XP_016862590.1 13 7000
XM_017007102.1 XP_016862591.1 11 6795
GenBank mRNA AK096450.1 - 1 100


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BF797567.1 - 1 61 Chr. 3 - 141944719 - 141947352 [-]
GenBankmRNA BM464287.1 - 352 Chr. - - [-]
GenBankmRNA DA766812.1 - 1 100 Chr. 3 - 141946820 - 141947391 [-]
GenBankmRNA BU596890.1 - 1 44 Chr. 3 - 141946601 - 141947333 [-]
GenBankmRNA HY114495.1 - 1 99 Chr. 3 - 141946999 - 141947390 [-]
GenBankmRNA DA233188.1 - 2 353 Chr. 3 - 141947046 - 141948656 [-]
GenBankmRNA DB135683.1 - 1 135 Chr. 3 - 141946975 - 141947426 [-]
GenBankmRNA HY104526.1 - 1 224 Chr. 3 - 141947053 - 141947515 [-]
GenBankmRNA DB155360.1 - 1 228 Chr. 3 - 141946937 - 141947506 [-]

View Full Product Details