Gene Transcripts
- Gene Symbol
- FFAR1
- Entrez Gene ID
- 2864
- Gene Name
- free fatty acid receptor 1
- Gene Aliases
- FFA1R, GPCR40, GPR40
- Chromosome Location
- Chr. 19: 35351542 - 35352464 on Build GRCh38
- UniGene ID
- Hs.248127
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC039436.1 |
- |
1 |
628 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI603605.1 |
- |
1 |
634 |
|
Chr. 19 - 35352419 - 35353229 [+] |
View Full Product Details