Gene Transcripts
- Gene Symbol
- DDIAS
- Entrez Gene ID
- 220042
- Gene Name
- DNA damage induced apoptosis suppressor
- Gene Aliases
- C11orf82, noxin
- Chromosome Location
- Chr. 11: 82901695 - 82934659 on Build GRCh38
- UniGene ID
- Hs.165607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK058145.1 |
- |
8 |
1090 |
|
View Full Product Details