Gene Transcripts
- Gene Symbol
- NFX1
- Entrez Gene ID
- 4799
- Gene Name
- nuclear transcription factor, X-box binding 1
- Gene Aliases
- NFX2, TEG-42, Tex42
- Chromosome Location
- Chr. 9: 33290417 - 33371163 on Build GRCh38
- UniGene ID
- Hs.413074
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055847.1 |
- |
1 |
17 |
|
View Full Product Details