Gene Transcripts
- Gene Symbol
- Baz2b
- Entrez Gene ID
- 407823
- Gene Name
- bromodomain adjacent to zinc finger domain, 2B
- Gene Aliases
- 5830435C13Rik, BC053917, D2Ertd794e
- Chromosome Location
- Chr. 2: 59899363 - 60209846 on Build GRCm38
- UniGene ID
- Mm.228071
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK079809.1 |
- |
3 |
357 |
|
AK166079.1 |
- |
3 |
345 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB635531.1 |
- |
3 |
357 |
|
Chr. 2 - 60206152 - 60209801 [-] |
GenBankmRNA |
BY755683.1 |
- |
3 |
345 |
|
Chr. 2 - 60206135 - 60209788 [-] |
GenBankmRNA |
BY734403.1 |
- |
3 |
337 |
|
Chr. 2 - 60206155 - 60209781 [-] |
View Full Product Details