Gene Transcripts

Gene Symbol
Baz2b
Entrez Gene ID
407823
Gene Name
bromodomain adjacent to zinc finger domain, 2B
Gene Aliases
5830435C13Rik, BC053917, D2Ertd794e
Chromosome Location
Chr. 2: 59899363 - 60209846 on Build GRCm38
UniGene ID
Mm.228071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK079809.1 - 3 382
AK166079.1 - 3 370


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB635531.1 - 3 382 Chr. 2 - 60206152 - 60209801 [-]
GenBankmRNA BY755683.1 - 3 370 Chr. 2 - 60206135 - 60209788 [-]
GenBankmRNA BY734403.1 - 3 362 Chr. 2 - 60206155 - 60209781 [-]

View Full Product Details