Gene Transcripts
- Gene Symbol
- Ggcx
- Entrez Gene ID
- 56316
- Gene Name
- gamma-glutamyl carboxylase
- Gene Aliases
- Chromosome Location
- Chr. 6: 72414296 - 72431106 on Build GRCm38
- UniGene ID
- Mm.19937
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK030779.1 |
- |
|
1370 |
|
View Full Product Details