Gene Transcripts
- Gene Symbol
- Klrb1-ps1
- Entrez Gene ID
- 724020
- Gene Name
- killer cell lectin-like receptor subfamily B member 1, pseudogene 1
- Gene Aliases
- Nkrp1e
- Chromosome Location
- Chr. 6: 129116518 - 129129446 on Build GRCm38
- UniGene ID
- Mm.424965
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073569.1 |
|
5 |
485 |
|
GenBank mRNA |
DQ237930.1 |
- |
5 |
485 |
|
View Full Product Details