Gene Transcripts
- Gene Symbol
- Umps
- Entrez Gene ID
- 22247
- Gene Name
- uridine monophosphate synthetase
- Gene Aliases
- 1700095D23Rik, AA408257, AL033308, BB164745
- Chromosome Location
- Chr. 16: 33954784 - 33967302 on Build GRCm38
- UniGene ID
- Mm.13145
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007077.1 |
- |
2 |
699 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707148.1 |
- |
2 |
698 |
|
Chr. 16 - 33966239 - 33967312 [+] |
View Full Product Details