Gene Transcripts
- Gene Symbol
- Hnrnpr
- Entrez Gene ID
- 74326
- Gene Name
- heterogeneous nuclear ribonucleoprotein R
- Gene Aliases
- 2610003J05Rik, 2610528B01Rik, Hnrpr
- Chromosome Location
- Chr. 4: 136310942 - 136345981 on Build GRCm38
- UniGene ID
- Mm.31051
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012160.1 |
- |
1 |
325 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM948891.1 |
- |
1 |
230 |
|
Chr. 4 - 136344805 - 136345563 [+] |
View Full Product Details