Gene Transcripts
- Gene Symbol
- Dhh
- Entrez Gene ID
- 13363
- Gene Name
- desert hedgehog
- Gene Aliases
- C78960
- Chromosome Location
- Chr. 15: 98881630 - 98898572 on Build GRCm38
- UniGene ID
- Mm.324688
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK134149.1 |
- |
1 |
52 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY031217.1 |
- |
1 |
52 |
|
Chr. 15 - 98893351 - 98894164 [+] |
View Full Product Details