Gene Transcripts
- Gene Symbol
- Synpo2
- Entrez Gene ID
- 118449
- Gene Name
- synaptopodin 2
- Gene Aliases
- 1110069I04Rik, 2310068J10Rik, 9530006G20Rik, AI848603, Myo
- Chromosome Location
- Chr. 3: 123076519 - 123236169 on Build GRCm38
- UniGene ID
- Mm.34359
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK004418.1 |
- |
1 |
110 |
|
View Full Product Details