Gene Transcripts
- Gene Symbol
- St3gal2
- Entrez Gene ID
- 20444
- Gene Name
- ST3 beta-galactoside alpha-2,3-sialyltransferase 2
- Gene Aliases
- AI429591, AW822065, ST3GalII, Siat5
- Chromosome Location
- Chr. 8: 110919833 - 110972497 on Build GRCm38
- UniGene ID
- Mm.200388
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK037177.1 |
- |
|
638 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE654362.1 |
- |
1 |
249 |
|
Chr. 8 - 110972805 - 110973589 [+] |
View Full Product Details