Gene Transcripts
- Gene Symbol
- Fbxw11
- Entrez Gene ID
- 103583
- Gene Name
- F-box and WD-40 domain protein 11
- Gene Aliases
- 2310065A07Rik, AA536858, BTRC2, BTRCP2, Fbxw1b, HOS
- Chromosome Location
- Chr. 11: 32642003 - 32746816 on Build GRCm38
- UniGene ID
- Mm.28017
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK010033.1 |
- |
1 |
447 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB730861.1 |
- |
1 |
206 |
|
Chr. 11 - 32680371 - 32680831 [+] |
View Full Product Details