Gene Transcripts
- Gene Symbol
- Zc3h12c
- Entrez Gene ID
- 244871
- Gene Name
- zinc finger CCCH type containing 12C
- Gene Aliases
- A230108E06, C230027N18Rik, mKIAA1726
- Chromosome Location
- Chr. 9: 52109583 - 52169154 on Build GRCm38
- UniGene ID
- Mm.295686
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042784.1 |
- |
3 |
323 |
|
View Full Product Details