Gene Transcripts
- Gene Symbol
- Klhl38
- Entrez Gene ID
- 268807
- Gene Name
- kelch-like 38
- Gene Aliases
- 8230402K04Rik
- Chromosome Location
- Chr. 15: 58314573 - 58324169 on Build GRCm38
- UniGene ID
- Mm.124498
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042526.1 |
- |
|
647 |
|
View Full Product Details