Gene Transcripts
- Gene Symbol
- Dis3l2
- Entrez Gene ID
- 208718
- Gene Name
- DIS3 mitotic control homolog (S. cerevisiae)-like 2
- Gene Aliases
- 4930429A22Rik, 8030493P09Rik
- Chromosome Location
- Chr. 1: 86703779 - 87050097 on Build GRCm38
- UniGene ID
- Mm.389152
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020223.1 |
- |
1 |
563 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY720465.1 |
- |
1 |
563 |
|
Chr. 1 - 86857371 - 86858005 [+] |
View Full Product Details