Gene Transcripts
- Gene Symbol
- Slit1
- Entrez Gene ID
- 20562
- Gene Name
- slit homolog 1 (Drosophila)
- Gene Aliases
- Slil1, mKIAA0813
- Chromosome Location
- Chr. 19: 41597824 - 41747275 on Build GRCm38
- UniGene ID
- Mm.39155
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK049808.1 |
- |
1 |
380 |
|
View Full Product Details