Gene Transcripts
- Gene Symbol
- Sh3glb1
- Entrez Gene ID
- 54673
- Gene Name
- SH3-domain GRB2-like B1 (endophilin)
- Gene Aliases
- AA409932, AI314629, AU015566, Bif-1, mKIAA0491
- Chromosome Location
- Chr. 3: 144683678 - 144720398 on Build GRCm38
- UniGene ID
- Mm.271775
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK052668.1 |
- |
1 |
937 |
|
AK086477.1 |
- |
1 |
877 |
|
AK147816.1 |
- |
1 |
914 |
|
View Full Product Details