Gene Transcripts
- Gene Symbol
- Slc38a9
- Entrez Gene ID
- 268706
- Gene Name
- solute carrier family 38, member 9
- Gene Aliases
- 4833412L08Rik, 6720411P22Rik, 9130023D20Rik, 9430067K09Rik, A730092C09
- Chromosome Location
- Chr. 13: 112660691 - 112738752 on Build GRCm38
- UniGene ID
- Mm.397106
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020109.1 |
- |
1 |
492 |
|
View Full Product Details