Gene Transcripts
- Gene Symbol
- Slc7a5
- Entrez Gene ID
- 20539
- Gene Name
- solute carrier family 7 (cationic amino acid transporter, y+ system), member 5
- Gene Aliases
- 4F2LC, D0H16S474E, LAT1, TA1
- Chromosome Location
- Chr. 8: 121881146 - 121907686 on Build GRCm38
- UniGene ID
- Mm.22374
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK183612.1 |
- |
2, 3 |
120 |
|
AK189161.1 |
- |
2, 3 |
120 |
|
View Full Product Details