Gene Transcripts
- Gene Symbol
- Wwp1
- Entrez Gene ID
- 107568
- Gene Name
- WW domain containing E3 ubiquitin protein ligase 1
- Gene Aliases
- 8030445B08Rik, AIP5, SDRP1, Tiul1
- Chromosome Location
- Chr. 4: 19608296 - 19709004 on Build GRCm38
- UniGene ID
- Mm.474845
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI847908.1 |
- |
1 |
173 |
|
AK038924.1 |
- |
|
718 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB178800.2 |
- |
1 |
170 |
|
Chr. 4 - 19607796 - 19608409 [+] |
View Full Product Details