Gene Transcripts
- Gene Symbol
- Rhox13
- Entrez Gene ID
- 73614
- Gene Name
- reproductive homeobox 13
- Gene Aliases
- 1700123J19Rik
- Chromosome Location
- Chr. X: 38120840 - 38129966 on Build GRCm38
- UniGene ID
- Mm.50505
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007253.1 |
- |
3 |
211 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707355.1 |
- |
3 |
195 |
|
Chr. X - 38121714 - 38129966 [+] |
GenBankmRNA |
CF356891.1 |
- |
3 |
165 |
|
Chr. X - 38121744 - 38129972 [+] |
View Full Product Details