Gene Transcripts
- Gene Symbol
- Med26
- Entrez Gene ID
- 70625
- Gene Name
- mediator complex subunit 26
- Gene Aliases
- 5730493L18Rik, AI414941, AW495270, Crsp7
- Chromosome Location
- Chr. 8: 72493453 - 72548310 on Build GRCm38
- UniGene ID
- Mm.235885
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042883.1 |
- |
|
51 |
|
AK054023.1 |
- |
|
41 |
|
- Gene Symbol
- LOC102639683
- Entrez Gene ID
- 102639683
- Gene Name
- uncharacterized LOC102639683
- Gene Aliases
- Chromosome Location
- Chr. 8: 72492731 - 72504592 on Build GRCm38
- UniGene ID
- Mm.440960
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK085371.1 |
- |
|
51 |
|
View Full Product Details