Gene Transcripts
- Gene Symbol
- Has2
- Entrez Gene ID
- 15117
- Gene Name
- hyaluronan synthase 2
- Gene Aliases
- Chromosome Location
- Chr. 15: 56665627 - 56694546 on Build GRCm38
- UniGene ID
- Mm.5148
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK045020.1 |
- |
1 |
2566 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB293662.2 |
- |
1 |
488 |
|
Chr. 15 - 56691948 - 56692461 [-] |
View Full Product Details