Gene Transcripts
- Gene Symbol
- Wdr66
- Entrez Gene ID
- 269701
- Gene Name
- WD repeat domain 66
- Gene Aliases
- 4930415N18Rik, 4933428F06Rik, 5031404N07
- Chromosome Location
- Chr. 5: 123254560 - 123327484 on Build GRCm38
- UniGene ID
- Mm.444012
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015152.1 |
- |
1 |
662 |
|
View Full Product Details