Gene Transcripts
- Gene Symbol
- Gmnn
- Entrez Gene ID
- 57441
- Gene Name
- geminin
- Gene Aliases
- AW546347, Gem
- Chromosome Location
- Chr. 13: 24751845 - 24761937 on Build GRCm38
- UniGene ID
- Mm.12239
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131677.1 |
- |
1 |
579 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV124533.2 |
- |
1 |
146 |
|
Chr. 13 - 24762018 - 24762197 [+] |
View Full Product Details