Gene Transcripts
- Gene Symbol
- Ints7
- Entrez Gene ID
- 77065
- Gene Name
- integrator complex subunit 7
- Gene Aliases
- 5930412E23Rik, int7
- Chromosome Location
- Chr. 1: 191575641 - 191622676 on Build GRCm38
- UniGene ID
- Mm.338175
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020029.1 |
- |
|
1327 |
|
View Full Product Details