Gene Transcripts
- Gene Symbol
- Gatsl2
- Entrez Gene ID
- 80909
- Gene Name
- GATS protein-like 2
- Gene Aliases
- 7530428J21Rik, AW742413, Gats, Gatsl1
- Chromosome Location
- Chr. 5: 134099748 - 134141782 on Build GRCm38
- UniGene ID
- Mm.426767
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033062.1 |
- |
|
1301 |
|
AK035431.1 |
- |
|
1317 |
|
BC026208.1 |
- |
|
702 |
|
View Full Product Details