Gene Transcripts
- Gene Symbol
- Lemd1
- Entrez Gene ID
- 213409
- Gene Name
- LEM domain containing 1
- Gene Aliases
- 4930540I23Rik, 6430514M23Rik, AI428128, LEMP-1
- Chromosome Location
- Chr. 1: 132190687 - 132257382 on Build GRCm38
- UniGene ID
- Mm.426321
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK032278.1 |
- |
1 |
1522 |
|
BC066109.1 |
- |
2 |
2640 |
|
View Full Product Details