Gene Transcripts
- Gene Symbol
- GUSBP11
- Entrez Gene ID
- 91316
- Gene Name
- glucuronidase, beta pseudogene 11
- Gene Aliases
- Chromosome Location
- Chr. 22: 23638487 - 23717423 on Build GRCh38
- UniGene ID
- Hs.148656
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000445682.2 |
- |
11, 12 |
2899 |
|
Chr. 22 - 23638487 - 23717356 [-] |
GENCODE |
ENST00000455485.5 |
- |
4, 5 |
3365 |
|
Chr. 22 - 23638487 - 23693151 [-] |
GENCODE |
ENST00000422506.5 |
- |
9, 10 |
1604 |
|
Chr. 22 - 23638487 - 23717356 [-] |
GENCODE |
ENST00000452737.5 |
- |
9, 10 |
1211 |
|
Chr. 22 - 23638487 - 23717321 [-] |
View Full Product Details