Gene Transcripts
- Gene Symbol
- PSMD6-AS2
- Entrez Gene ID
- 100507062
- Gene Name
- PSMD6 antisense RNA 2
- Gene Aliases
- Chromosome Location
- Chr. 3: 64004022 - 64012243 on Build GRCh38
- UniGene ID
- Hs.152536
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038286.1 |
|
1 |
1162 |
|
GenBank mRNA |
AK023371.1 |
- |
1 |
1162 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000472046.1 |
- |
1 |
1162 |
|
Chr. 3 - 64004022 - 64012148 [+] |
View Full Product Details