Gene Transcripts
- Gene Symbol
- GUSBP11
- Entrez Gene ID
- 91316
- Gene Name
- glucuronidase, beta pseudogene 11
- Gene Aliases
- Chromosome Location
- Chr. 22: 23638487 - 23717423 on Build GRCh38
- UniGene ID
- Hs.148656
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538181.1 |
- |
11, 12 |
2859 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000434893.1 |
- |
2, 3 |
225 |
|
Chr. 22 - 23653170 - 23686914 [-] |
GENCODE |
ENST00000421064.5 |
- |
11, 12 |
2862 |
|
Chr. 22 - 23653169 - 23717347 [-] |
View Full Product Details