Gene Transcripts
- Gene Symbol
- 2010204K13Rik
- Entrez Gene ID
- 68355
- Gene Name
- RIKEN cDNA 2010204K13 gene
- Gene Aliases
- 0610010M13Rik, Cip7
- Chromosome Location
- Chr. X: 7411817 - 7422988 on Build GRCm38
- UniGene ID
- Mm.45111
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027924.1 |
|
2 |
712 |
|
GenBank mRNA |
AK077356.1 |
- |
2 |
758 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB027336.2 |
- |
1 |
558 |
|
Chr. X - 7411817 - 7412486 [-] |
View Full Product Details